Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone Safety White SMS Lab Coats, Snap Front, Elastic Wrists
SDP

Catalog No. 19168636
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-168-636 Small
19-168-637 Medium
19-168-639 X-Large
19-168-638 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-168-636 Supplier Keystone Safety Supplier No. LC0WESMSSMWHITE
This item is not returnable. View return policy

Latex free SMS lab coats provide barrier protection and are comfortable to wear.

  • Latex free, 3-layer SMS fabric
  • Barrier against water based liquids and light chemical splash
  • Single collar
  • Strong, high tensile strength
  • Comfortable, lightweight, breathable
  • Serged seams
  • Meets ANSI 101-1996 and Federal Flammability ACT 16 CFR 1610.
  • Multi use for cleanroom, pharmaceutical, food, janitorial, cleaning, maintenance
  • Keystone LC0-WE-SMS-WHITE SMS Lab Coat, No Pockets, Elastic Wrists, Snap Front
  • White, Pack of 30

Specifications

Product Type Controlled Environments Lab Coat
Size Small
Gender Unisex
Material SMS
Color White
Closure Type Snaps
Cuff Style Elastic
Disposable Single-use
Sterility Non-sterile
No. of Pockets None
Collar Type Single
Latex Free Yes
Model LC0-WE-SMS
Seam Type Serged
Certifications/Compliance ANSI 101-1996
For Use With (Application) Janitorial, Manufacturing, Food Service, Industrial, Medical, Cleanroom, Pharmaceutical
Length (English) 38.5 in.
Length (Metric) 97.79 cm
Weight (English) 1.4 oz.
Weight (Metric) 40 g
Autoclavable Not Autoclavable
No. per Pack 30/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.