Learn More
Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| pH | 2.1 |
| Melting Point | 8°C |
| Boiling Point | 101°C |
| Color | Colorless |
| Linear Formula | HCOOH |
| UN Number | 1779 |
| Formula Weight | 46.03 |
| Grade | Laboratory |
| Density | 1.220g/cm³ |
| Quantity | 1 L |
| Show More |
Safety and Handling
| Hazard Statement | DANGER! |
| Precautionary Statement | Emergency Overview Flammable liquid and vapor. Causes severe burns by all exposure routes. Hygroscopic. Wear personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen. NFPA Health:3 Flammability:2 Instability:0 |
| DOTInformation | DOT Class 8, : Corrosive |
| Recommended Storage | Store separately from sulfuric acid and caustics. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
