Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Phosphoric Acid, 85% (HPLC), Fisher Chemical

Catalog No. A260500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Flint Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A260-500 500 mL Flint Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. A260-500
Only null left
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 7664-38-2 , 7732-18-5
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
MDL Number MFCD00011340
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O

Specifications

pH 1
Melting Point 21°C
Boiling Point 158°C
Density 1.680g/cm³
Heavy Metals (as Pb) 0.001% max.
Identification Pass Test
Packaging Flint Glass Bottle
Reducing Substances Pass Test
Vapor Pressure 2 hPa at 20°C
Assay Percent Range ≥85.0 %
Quantity 500 mL
Physical Form Liquid
UN Number 1805
Formula Weight 98
Percent Purity ≥85.0%
Grade Also meets ACS specifications
Chemical Name or Material o-Phosphoric Acid, 85%
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
DOTInformation Class 8: Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.