Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Phosphoric Acid (Cryst. or Flakes/Certified ACS), Fisher Chemical™
GSA_VA

Catalog No. A280100 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
3 kg
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A280-100 100 g Glass Bottle
A280-500 500 g Glass Bottle
A280-3 3 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A280-100
Only null left
Add to Cart

Mixture of meta-Phosphoric Acid and Sodium Metaphosphate

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37267-86-0 , 50813-16-6
Molecular Formula HO3P
Molecular Weight (g/mol) 79.979
MDL Number MFCD00011351
InChI Key UEZVMMHDMIWARA-UHFFFAOYSA-N
Synonym metaphosphoric acid, meta-phosphoric acid, hydrogenphosphonate, m-phosphoric acid, phosphenic acid, unii-mtk99r3uv0, metaphosphoric acid hpo3, x-phosphate-group, phosphonate 1-, hydrogen phosphonate
PubChem CID 3084658
SMILES OP(=O)=O

Specifications

Density 2.4g/cm³
Heavy Metals (as Pb) 0.005% max.
Identification Pass Test
Packaging Glass Bottle
Assay Percent Range 33.5 to 36.5 %
Quantity 100 g
Physical Form Solid
Percent Purity 33.5 to 36.5%
Grade Certified ACS
Chemical Name or Material m-Phosphoric Acid
Safety handling Solvent Safety handling Check Alert storage code Red
DOTInformation DOT Class 8, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.