Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid (ACS Grade), Fisher Chemical™

Catalog No. A300S500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
6.5 gal.
6 x 2.5 L
225 lb.
700 lb.
Packaging:
Glass Bottle
PPQ Resin Bottle
Poly Bottle
Poly Drum
Safe-Cote™ Glass Bottle
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A300S-500 500 mL Safe-Cote™ Glass Bottle
A300-500 500 mL Glass Bottle
A300-212 2.5 L Poly Bottle
A300C-212 2.5 L PPQ Resin Bottle
A300SI-212 2.5 L Safe-Cote™ Glass Bottle
A300S-212 6 x 2.5 L Safe-Cote™ Glass Bottle
A300-612GAL 6.5 gal. Glass Bottle
A300-225LB 225 lb. Poly Drum
A300-700LB 700 lb. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. A300S-500
  • $219.00
    $160.65 / Each

    Save $58.35

    27% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $1,008.00
    $755.65 / Case of 6 EA

    Save $252.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Approx. Normality is 36

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

pH 0.3
Color Colorless to Brown
Melting Point 10°C
Boiling Point 290°C
Density 1.84g/cm³
Heavy Metals (as Pb) 0.8ppm max.
Identification Pass Test
Packaging Safe-Cote™ Glass Bottle
Residue after Ignition 5 ppm max.
Vapor Pressure <0.001 mmHg at 20°C
Normality about 36
Assay Percent Range 95.0 to 98.0 w/w %
Quantity 500 mL
Physical Form Liquid
UN Number 1830
Formula Weight 98.08
Percent Purity 95.0 to 98.0% (w/w)
Grade ACS Grade
Chemical Name or Material Sulfuric Acid
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Causes severe burns by all exposure routes. Reacts violently with water. Contact with combustible material may cause fire. Exposure to strong inorganic mists containing sulfuric acid may cause cancer by inhalation. Hygroscopic. Use personal protective equipment. Do not allow contact with water. Keep away from clothing and other combustible materials. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required.

NFPA

Health:3
Flammability:0
Instability:2
DOTInformation DOT Class 8, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.