Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol, CDA 19 (Histological), Fisher Chemical™
GSA_VA

Catalog No. A406F1GAL Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 gal.
4 L
20 L
Packaging:
Lined Steel Drum
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A406F-1GAL 1 gal. Poly Bottle
A406P-4 4 L Poly Bottle
A406-20 20 L Lined Steel Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A406F-1GAL
Only null left
Add to Cart

Nonvolatile matter, ≤0.003%

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5 , 108-10-1 , 108-88-3 , 110-54-3 , 7732-18-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
MDL Number MFCD00003568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -85°C
Density 0.813g/cm³
Quantity 1 gal.
Identification Pass Test
Packaging Poly Bottle
Physical Form Liquid
Vapor Pressure 50mmHg at 20°C
UN Number 1170
Boiling Point 79°C
Grade Histological
Chemical Name or Material Ethanol, CDA 19
Show More Show Less
DOTInformation DOT Class 3, : Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.