Learn More
Specifications
Specifications
| Melting Point | -98°C |
| Color | Colorless |
| Density | 0.791g/cm³ |
| Quantity | 500 mL |
| Flash Point | 12°C (53.6°F) |
| Identification | Pass test |
| Packaging | HDPE Bottle |
| Physical Form | Liquid |
| Residue after Evaporation | 10ppm max. |
| Specific Gravity | 0.791 |
| Show More |
Safety and Handling
| Hazard Category | DANGER! |
| Hazard Statement | Emergency Overview Flammable liquid and vapor. Cannot be made non-poisonous. Vapor harmful. Toxic by inhalation, in contact with skin and if swallowed. Irritating to eyes and skin. May cause irritation of respiratory tract. Toxic: danger of very serious irreversible effects through inhalation, in contact with skin and if s. Explosive. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. NFPA Health:1 Flammability:3 Instability:0 |
| Precautionary Statement | Emergency Overview Flammable liquid and vapor. Cannot be made non-poisonous. Vapor harmful. Toxic by inhalation, in contact with skin and if swallowed. Irritating to eyes and skin. May cause irritation of respiratory tract. Toxic: danger of ve |
| DOTInformation | DOT Class 3, : Flammable Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.