Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol (HPLC), Fisher Chemical™

Catalog No. A4524 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
4 L
19 L
Packaging:
Glass Bottle
NOWPak™ I
Safe-Cote™ Amber Glass 
Wide Neck Amber Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A452-4 4 L Glass Bottle
A452-1 1 L Wide Neck Amber Bottle
A452SK-4 4 L Safe-Cote™ Amber Glass 
A452N1-19 19 L NOWPak™ I
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. A452-4
Get 1 Extra Chemical When You Buy 2
View all promotions
Add to Cart

HPLC mobile phase for analytical and preparative separations

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Absorbance 0.025 AU max. at 254nm, 0.15 AU max. at 230nm, 0.30 AU max. at 220nm, 1.00 AU max. at 205nm
Color Colorless
Quantity 4 L
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 3ppm max.
Specific Gravity 0.791
Assay Percent Range Assay: 99.9% min by GC
Titratable Acid 0.0003mEq/g max.
Titratable Base 0.0002mEq/g max.
Vapor Pressure 128 hPa at 20°C
Linear Formula CH3OH
Viscosity 0.55 cP at 20°C
UN Number 1230
Water 0.1% max.
Boiling Point 64.7°C
Solubility Information Solubility in water: miscible
Formula Weight 32.04
Percent Purity ≥99.9%
Grade Mobile phase for HPLC applications
Chemical Name or Material Methanol
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Cannot be made non-poisonous. Vapor harmful. Toxic by inhalation, in contact with skin and if swallowed. Irritating to eyes and skin. May cause irritation of respiratory tract. Toxic: danger of very serious irreversible effects through inhalation, in contact with skin and eyes. Explosive. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required.

NFPA

Health:1
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.