Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methanol (Purge and Trap Suitable for Volatile Organic Residue Analysis), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Glass
Specifications
Specifications
| Melting Point | -98°C |
| Color | Colorless |
| Density | 0.791g/cm³ |
| Quantity | 500 mL |
| Flash Point | 12°C (53.6°F) |
| Packaging | Amber Glass |
| Physical Form | Liquid |
| Residue after Evaporation | 1ppm max. |
| Assay Percent Range | ≥99.9 % |
| Titratable Acid | 0.0003mEq/g max. |
| Show More |
Safety and Handling
| Hazard Statement | DANGER! |
| Precautionary Statement | Emergency Overview Flammable liquid and vapor. Cannot be made non-poisonous. Vapor harmful. Toxic by inhalation, in contact with skin and if swallowed. Irritating to eyes and skin. May cause irritation of respiratory tract. Toxic: danger of very serious irreversible effects through inhalation, in contact with skin and if s. Explosive. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. NFPA Health:1 Flammability:3 Instability:0 |
| DOTInformation | DOT Class 3, : Flammable Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction