Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perchloric Acid (Optima™), Fisher Chemical™

Catalog No. A4691 Shop All Fisherbrand Products
Click to view available options
Quantity:
250 mL
500 mL
1 L
Packaging:
FEP Bottle

Catalog No. A469-1

Only null left
Add to Cart
  • Ideal for analysis by ICP-MS and high-resolution ICP-MS.
  • Evaluated for 65 metals at low-level ppt concentrations.
  • Manufactured, purified, and packaged in a Class 1000 clean room.
  • Packaged in specially cleaned Teflon FEP for PFA bottles to maintain product integrity and to eliminate metal leaching from glass bottles.
  • Three year shelf life from time of manufacture - available on COA
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7601-90-3
Molecular Formula ClHO4
Molecular Weight (g/mol) 100.45
MDL Number MFCD00011325
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
PubChem CID 24247
ChEBI CHEBI:29221
IUPAC Name perchloric acid
SMILES O[Cl](=O)(=O)=O

Specifications

pH 0.1
Color Colorless
Melting Point -18°C
Boiling Point 203°C
Density 1.660g/cm³
Packaging FEP Bottle
Vapor Pressure 6.8mmHg at 25°C
Certificate of Analysis Certificate of Lot Analysis included with each bottle.
Quantity 1 L
Physical Form Liquid
UN Number 1873
Merck Index 15, 7267
Formula Weight 100.46
Grade OPTIMA
Chemical Name or Material Perchloric Acid
Show More Show Less
DOTInformation DOT Class 5.1 : Oxidizer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.