Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid (TraceMetal™ Grade), Fisher Chemical™
GSA_VA

Catalog No. A510P212 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
HDPE Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A510-P212 2.5 L HDPE Bottle
A510-P500 500 mL HDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A510-P212
Get Up to $1,800 in Chemicals with Purchase
View all promotions
 Restricted Item
Add to Cart

missing translation for 'validMainframeMsg'

  • Certificate of Lot Analysis included with each shipment.
  • Ideal for analysis by IC, GF or Flame AA, ICP-OES and ICP-MS.
  • Evaluated for 65 metals at <1 ppb concentrations. Manufactured, purified, and packaged in a Class1000 clean room.
  • Packaged in non-breakable polypropylene bottles to maintain product integrity and to eliminate metal leaching from glass bottles.
  • Three year shelf life from time of manufacture available on COA. 45mm bottle neck size.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

pH 0.3
Color Colorless to Brown
Melting Point 10°C
Boiling Point 290°C
Density 1.84g/cm³
Packaging HDPE Bottle
Vapor Pressure <0.001 mmHg at 20°C
Certificate of Analysis Certificate of Lot Analysis included with each shipment.
Quantity 2.5 L
Physical Form Liquid
UN Number 1830
Formula Weight 98.08
Grade TraceMetal
Chemical Name or Material Sulfuric Acid
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
DOTInformation DOT Class 8, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.