Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Hydroxide (Certified ACS Plus), Fisher Chemical™
GSA_VA

Catalog No. A669212 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
6 x 2.5 L
385 lb.
Packaging:
Glass Bottle
Poly Drum
Safe-Cote™ Glass
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A669-212 2.5 L Glass Bottle
A669-500 500 mL Glass Bottle
A669S-500 500 mL Safe-Cote™ Glass
A669C-212 6 x 2.5 L Glass Bottle
A669S-212 6 x 2.5 L Safe-Cote™ Glass
A669-385LB 385 lb. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. A669-212
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
This item is not returnable. View return policy

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1336-21-6 , 7664-41-7 , 7732-18-5
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name amine hydrate
SMILES N.O

Specifications

pH 12
Color Colorless
Boiling Point 38°C
Melting Point -57°C
Assay Percent Range Assay: 28.0 to 30.0 w/w %
Linear Formula NH4OH
Quantity 2.5 L
Formula Weight 35.05
Percent Purity 28.0 to 30.0% (w/w)
Grade Certified ACS Plus
Identification Pass test
Normality about 14.8N
Packaging Glass Bottle
Residue after Ignition 3ppm max.
Vapor Pressure 500 hPa at 20°C
Physical Form Liquid
Chemical Name or Material Ammonium Hydroxide, about 14.8N
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
DOTInformation DOT Class 8: Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.