Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol (Low Water), Fisher Chemical™

Catalog No. A9354 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A935-4 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A935-4
  • $280.50
    $178.65 / Each

    Save $101.85

    36% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $831.00
    $529.65 / Case of 4 EA

    Save $301.35

    36% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Low water concentration

  • Also available in recyclable FisherPak™ and NOWPak™ containers.
  • EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Color Colorless
Density 0.791g/cm³
Quantity 4 L
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 5ppm max.
Assay Percent Range ≥99.8 %
Titratable Acid 0.0003mEq/g max.
Titratable Base 0.0002mEq/g max.
Vapor Pressure 128 hPa at 20°C
Linear Formula CH3OH
Viscosity 0.55 cP at 20°C
Water 0.008% max.
Boiling Point 64.7°C
Formula Weight 32.04
Percent Purity ≥99.8%
Chemical Name or Material Methanol
Grade Also meets ACS specifications
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Category DANGER!
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Cannot be made non-poisonous. Vapor harmful. Toxic by inhalation, in contact with skin and if swallowed. Irritating to eyes and skin. May cause irritation of respiratory tract. Toxic: danger of very serious irreversible effects through inhalation, in contact with skin and if s. Explosive. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required.

NFPA

Health:1
Flammability:3
Instability:0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.