Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reagent Alcohol, Fisher Chemical™
GSA_VA

Catalog No. A9624 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 gal.
4 L
200 L
Packaging:
Aluminum Can
Glass Bottle
Poly Bottle
Steel Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A962-4 4 L Glass Bottle
A962P-4 4 L Poly Bottle
A962-200 200 L Steel Drum
A962F-1GAL 1 gal. Poly Bottle
A962S-4 4 L Aluminum Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. A962-4
  • $336.50
    $253.65 / Each

    Save $82.85

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $1,035.00
    $794.65 / Case of 4 EA

    Save $240.35

    23% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Specially Denatured Alcohol for histologic use.

Note:  Federal registration is required to purchase sizes greater than 4L.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5 , 67-63-0 , 67-56-1
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
MDL Number MFCD00003568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -114.1°C
Density 0.8g/cm³
Quantity 4 L
Flash Point 12.8°C (55°F)
Packaging Glass Bottle
Physical Form Liquid
Assay Percent Range Assay: Ethanol (88 to 91%), Methanol (4.0 to 5.0% v/v), Isopropyl Alcohol (4.5 to 5.5%)
Specific Gravity 0.782 to 0.793
Vapor Pressure 44mmHg
UN Number 1170
Water 0.10% Max.
Boiling Point 78.5°C
Grade Reagent
Concentration or Composition (by Analyte or Components) Anhydrous Ethyl Alcohol 90% ±1% v/v; Methyl Alcohol approx. 5% v/v; 2-Propanol approx. 5% v/v
Chemical Name or Material Reagent Alcohol
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Cannot be made non-poisonous. Harmful by inhalation, in contact with skin and if swallowed. Vapor harmful. Irritating to eyes and skin. May cause irritation of respiratory tract. May cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. This substance has caused adverse reproductive and fetal effects in humans. Substances known to cause developmental toxicity in humans. Danger of very serious irreversible effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Wear personal protective equipment.

NFPA

Health:2
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.