Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alcohol (HPLC, Meets ACS), Fisher Chemical™

Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
64-17-5, 67-56-1, 67-63-0
C2H6O
32.04
MFCD00004595
OKKJLVBELUTLKV-UHFFFAOYSA-N
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
702
CHEBI:16236
Ethanol
CO
64-17-5, 67-56-1, 67-63-0
C2H6O
32.04
MFCD00004595
OKKJLVBELUTLKV-UHFFFAOYSA-N
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
702
CHEBI:16236
Ethanol
CO
Melting Point -114.1°C
Absorbance 0.04 AU max. at 254nm, 0.35 AU max. at 220nm, 0.65 AU max. at 210nm, 1 AU max. at 205nm
Assay Percent Range 99.8% min by GC
Linear Formula CH3CH2OH
UN Number 1987; Clear
Grade Mobile phase for HPLC applications
Concentration or Composition (by Analyte or Components) Anhydrous Ethyl Alcohol 90% ±1% v/v; Methyl Alcohol approx. 5% v/v; 2-Propanol approx. 5% v/v
Density 1.24 (Air= 1.0)
Boiling Point 78.5°C
Quantity 4 L
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3580 to 1.3610 (at 25°C)
Residue after Evaporation 5ppm max.
Titratable Acid 0.0003mEq/g max.
Titratable Base 0.0002mEq/g max.
Vapor Pressure 44mmHg
Water 0.1% max.
Chemical Name or Material Alcohol
Show More Show Less

DANGER!

Emergency Overview
Flammable liquid and vapor. Cannot be made non-poisonous. Harmful by inhalation, in contact with skin and if swallowed. Vapor harmful. Irritating to eyes and skin. May cause irritation of respiratory tract. May cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. This substance has caused adverse reproductive and fetal effects in humans. Substances known to cause developmental toxicity in humans. Danger of very serious irreversible effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Wear personal protective equipment.

NFPA

Health:2
Flammability:3
Instability:0

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit