Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, Optima™, for HPLC and GC, Fisher Chemical™

Catalog No. A9961 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Soda Lime Glass
Safe-Cote™ Amber Glass 
Wide Neck Amber Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A996-1 1 L Wide Neck Amber Bottle
A996-4 4 L Amber Soda Lime Glass
A996SK-4 4 L Safe-Cote™ Amber Glass 
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A996-1
  • $346.00
    $175.65 / Each

    Save $170.35

    49% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $1,595.00
    $809.65 / Case of 6 EA

    Save $785.35

    49% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

High purity and versatile solvent suitable for HPLC, spectrophotometry, and pesticide residue analysis

  • Screened for fluorescing contaminants, and UV-absorbing contaminants in the 190-254nm range, to reduce interferences for smoother baselines
  • Tested for HPLC gradient suitability
  • Tested to ensure minimum halogenated residue levels (≤10 ng/L) to demonstrate suitability for pesticide analysis
  • Also meets ACS specifications
  • Intended for research use only
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -46°C
Absorbance 0.005 AU max. at 254nm, 0.02 AU max. at 220nm, 0.04 AU max. at 210nm, 0.05 AU max. at 205nm, 0.07 AU max. at 200nm, 0.15 AU max. at 195nm, 1.00 AU max. at 190nm
Color Colorless
Decomposition Information >500°C
Density 0.781g/cm³
Quantity 1 L
Flash Point 49°C (120.2°F)
Identification Pass Test
Packaging Wide Neck Amber Bottle
Physical Form Liquid
Refractive Index 1.3405 to 1.3425 (at 25°C)
Residue after Evaporation 0.8ppm max.
Assay Percent Range Assay: 99.9% min by GC
Titratable Acid 0.008mEq/g max.
Titratable Base 0.0006mEq/g max.
Vapor Pressure 97mbar at 20°C
Viscosity 0.36 cP at 20°C
Water 0.01% max.
UN Number 1648; Colorless
Boiling Point 81°C
Formula Weight 41.05
Percent Purity ≥99.9%
Grade High purity mobile phase for HPLC and/or extraction solvent for GC applications
Chemical Name or Material Acetonitrile
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Irritating to eyes. May cause skin and respiratory tract irritation. May cause central nervous system effects. Liver and kidney injuries may occur. Use personal protective equipment. Use only under a chemical fume hood. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.