Learn More
Specifications
Specifications
| Melting Point | -46°C |
| Absorbance | 0.01 max. at 254nm, 0.02 max. at 220nm, 0.04 max. at 210nm, 0.05 max. at 205nm, 0.07 max. at 200nm, 0.15 max. at 195nm, 1 max. at 190nm, Pass Test |
| Color | Colorless |
| Decomposition Information | >500°C |
| Quantity | 1 L |
| Flash Point | 12.8°C (55°F) |
| Identification | Pass Test |
| Packaging | Wide Neck Amber Bottle |
| Physical Form | Liquid |
| Residue after Evaporation | 1.0ppm max. |
| Show More |
Safety and Handling
| Hazard Statement | DANGER! |
| Precautionary Statement | Emergency Overview Flammable liquid and vapor. Irritating to eyes. May cause skin and respiratory tract irritation. May cause central nervous system effects. Liver and kidney injuries may occur. Use personal protective equipment. Use only under a chemical fume hood. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Move to fresh air. NFPA Health:2 Flammability:3 Instability:0 |
| DOTInformation | DOT Class 3, : Flammable Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.