Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile (HPLC), Fisher Chemical

Catalog No. A998N119 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 L
19 L
Packaging:
Amber Glass
Amber Glass Bottle
NOWPak™ I
Safe-Cote™ Amber Glass Bottle
Wide Neck Amber Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A998N1-19 19 L NOWPak™ I
A998SK-1 1 L Safe-Cote™ Amber Glass Bottle
A998-1 1 L Wide Neck Amber Bottle
A998-4 4 L Amber Glass Bottle
A998SK-4 4 L Safe-Cote™ Amber Glass Bottle
A998-212 2.5 L Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. A998N1-19
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

HPLC mobile phase for analytical and preparative separations

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -46°C
Absorbance 0.01 max. at 254nm, 0.02 max. at 220nm, 0.04 max. at 210nm, 0.05 max. at 205nm, 0.07 max. at 200nm, 0.15 max. at 195nm, 1 max. at 190nm, Pass Test
Color Colorless
Decomposition Information >500°C
Quantity 19 L
Flash Point 12.8°C (55°F)
Identification Pass Test
Packaging NOWPak™ I
Physical Form Liquid
Residue after Evaporation 1.0ppm max.
Specific Gravity 0.781
Assay Percent Range Assay: 99.9% min by GC
Titratable Acid 0.008mEq/g max.
Titratable Base 0.0006mEq/g max.
Vapor Pressure 97mbar at 20°C
Linear Formula CH3CN
Viscosity 0.36 cP at 20°C
Water 0.01% max.
UN Number 1648; Colorless
Merck Index 15, 69
Boiling Point 81°C
Solubility Information Soluble in water
Formula Weight 41.05
Percent Purity ≥99.9%
Grade Mobile phase for HPLC applications
Chemical Name or Material Acetonitrile
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Irritating to eyes. May cause skin and respiratory tract irritation. May cause central nervous system effects. Liver and kidney injuries may occur. Use personal protective equipment. Use only under a chemical fume hood. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.