Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-methoxyethyl) ether, 99%, extra pure

Catalog No. AC125850025
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC125850025 2.5 L Glass bottle
AC125852500 250 mL Glass bottle
AC125850010 1 L Glass bottle
AC125850100 10 L Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC125850025 Supplier Thermo Scientific Chemicals Supplier No. 125850025
Only null left
Add to Cart

CAS: 111-96-6 | C6H14O3 | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-96-6
Molecular Formula C6H14O3
Molecular Weight (g/mol) 134.18
InChI Key SBZXBUIDTXKZTM-UHFFFAOYSA-N
Synonym diglyme, bis 2-methoxyethyl ether, 2-methoxyethyl ether, 1-methoxy-2-2-methoxyethoxy ethane, diethylene glycol dimethyl ether, poly-solv, di 2-methoxyethyl ether, dimethyl carbitol, 2,5,8-trioxanonane
PubChem CID 8150
ChEBI CHEBI:46784
IUPAC Name 1-methoxy-2-(2-methoxyethoxy)ethane
SMILES COCCOCCOC

Specifications

Melting Point -64.0°C
Density 0.9370g/mL
Boiling Point 162.0°C
Flash Point 51°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4070 to 1.4090
Linear Formula (CH3OCH2CH2)2O
Quantity 2.5 L
Beilstein 01,IV,2393
Fieser 01,255
Specific Gravity 0.937
Merck Index 15,3184
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol,ether,hydrocarbon solvents
Viscosity 2 mPa.s (20°C)
Formula Weight 134.18
Percent Purity 99%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Bis(2-methoxyethyl) ether
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May damage fertility or the unborn child.
Flammable liquid and vapor.
May form explosive peroxides.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Use personal protective equipment as required.
EINECSNumber 203-924-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.