Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium hydroxide, ca. 85%, for analysis, pellets

Catalog No. AC134060250
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
10 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
Plastic drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC134060250 25 kg Fibre drum
AC134062500 250 g Plastic bottle
AC134060010 1 kg Plastic bottle
AC134060025 2.5 kg Plastic bottle
AC134060100 10 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC134060250 Supplier Thermo Scientific Chemicals Supplier No. 134060250
Add to Cart

CAS: 1310-58-3 | HKO | 56.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
SMILES [OH-].[K+]

Specifications

Boiling Point 1320.0°C
Melting Point 360.0°C
Assay Percent Range ca. 85%
Linear Formula KOH
Fieser 01,935; 02,346; 03,238; 04,409; 05,557; 06,486; 07,303; 08,415; 09,387; 11,439; 12,411; 13,258
Merck Index 15, 7761
Solubility Information Solubility in water: 1120g/L (20°C). Other solubilities: 1780g/L water (100°C), soluble in ethanol (400g/L), soluble in ammonia, slightly soluble in ether
Quantity 25 kg
IUPAC Name potassium;hydroxide
Formula Weight 56.11
Percent Purity ∼85%
Grade Analytical
Packaging Fibre drum
Physical Form Pellet
Chemical Name or Material Potassium hydroxide, ca. 85 %
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.