Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cinnamyl alcohol, 98% trans
Click to view available options
Quantity:
5 g
100 g
250 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 31.0°C to 35.0°C |
| Color | White |
| Density | 1.0400g/mL |
| Boiling Point | 258.0°C |
| Flash Point | 126°C |
| Infrared Spectrum | Authentic |
| Packaging | Glass bottle |
| Linear Formula | C6H5CH=CHCH2OH |
| Quantity | 100 g |
| Beilstein | 06, 570 |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | GHS Signal Word: Warning |
| missing translation for 'healthHazard2' | GHS H Statement May cause an allergic skin reaction. |
| missing translation for 'healthHazard3' | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Contaminated work clothing should not be allowed out of the workplace. Avoid breathing dust/fume/gas/mist/vapors/spray. |
| EINECSNumber | 203-212-3 |
| RTECSNumber | GE2200000 |
| TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction