Learn More
Methanol, 99.9%, for analysis
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -98.0°C |
| Density | 0.7910g/mL |
| Quantity | 1 L |
| Flash Point | 12°C |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Refractive Index | 1.3280 to 1.3300 |
| Residue after Evaporation | 0.002% max. |
| Assay Percent Range | 99.9% |
| Specific Gravity | 0.791 |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Causes damage to organs. Highly flammable liquid and vapor. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Gently wash with plenty of soap and water. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment. |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.