Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, 99.9%, for analysis

Catalog No. AC176845000
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 L
25 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC176845000 500 mL
AC176840010 1 L
AC176840025 2.5 L
AC176840100 10 L
AC176840250 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC176845000 Supplier Thermo Scientific Chemicals Supplier No. 176845000
  • $52.00
    $44.65 / Each

    Save $7.35

    14% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $240.00
    $204.65 / Case of 6 EA

    Save $35.35

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 67-56-1 | CH4O | 32.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98.0°C
Density 0.7910g/mL
Quantity 500 mL
Flash Point 12°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3280 to 1.3300
Residue after Evaporation 0.002% max.
Specific Gravity 0.791
Assay Percent Range 99.9%
Titratable Acid 0.0003 meq/g max.
Titratable Base 0.0002 meq/g max.
Linear Formula CH3OH
Viscosity 0.55 mPa.s (20°C)
Water 0.05% max. (K.F.)
Boiling Point 64.7°C
Solubility Information Solubility in water: miscible with water. Other solubilities: miscible with ethanol, ether, benzene, ketones and, most other organic solvents
Formula Weight 32.04
Percent Purity 99.9%
Grade Analytical
Chemical Name or Material Methanol
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.