Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium cerium(IV) nitrate, pure

Catalog No. AC212360010
Click to view available options
Quantity:
1 kg
5 kg

Catalog No. AC212360010

Supplier: Thermo Scientific Chemicals 212360010

Only null left
Add to Cart

Oxidation reagent | CAS: 16774-21-3 | H8CeN8O18 | 548.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16774-21-3
Molecular Formula H8CeN8O18
Molecular Weight (g/mol) 548.22
Synonym Ceric ammonium nitrate, CAN

Specifications

Melting Point 107.0°C to 108.0°C
Quantity 1 kg
Linear Formula (NH4)2Ce(NO3)6
Fieser 01,120; 02,63; 03,44; 04,71; 05,101; 06,99; 09,99; 10,79; 15,70; 16,66; 17,68
Merck Index 15,1991
Solubility Information Solubility in water: 1410g/L (20°C). Other solubilities: insoluble in fat, soluble in alcohol
Formula Weight 548.22
Grade Pure
Loss on Drying 2% max.
Chemical Name or Material Ammonium cerium(IV) nitrate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May intensify fire; oxidizer.
May be corrosive to metals.
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
EINECSNumber 240-827-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.