Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Pentane, 99+%, extra pure, anhydrous

Catalog No. AC217240251
Click to view available options
Quantity:
1 L
2.5 L
4 L
25 L

Catalog No. AC217240251

Supplier: Thermo Scientific Chemicals 217240251

Only null left
Add to Cart

CAS: 109-66-0 | C5H12 | 72.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC

Specifications

Melting Point -130.0°C
CAS Min % 99.0
CAS Max % 100.0
Density 0.6260g/mL
Boiling Point 36.0°C
Quantity 25 L
Flash Point −49°C
Infrared Spectrum Authentic
Packaging Metal Drum
Physical Form Liquid
Refractive Index 1.358
Assay Percent Range 99+%
Residue after Evaporation 0.001% max.
Specific Gravity 0.626
Linear Formula CH3(CH2)3CH3
Vapor Pressure Vapor Pressure: 573mbar at 20°C
Viscosity 0.25 mPa.s (20°C)
Beilstein 01, 130
Water 0.005% max.
Merck Index 15, 7228
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and most other common organic, solvents
Formula Weight 72.15
Percent Purity ≥99%
Grade Extra Pure
Chemical Name or Material n-Pentane, p.a.
Show More Show Less
Hazard Category Danger
Hazard Statement GHS P Statement May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor. Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.