Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic acid, ammonium salt, 98+%, for analysis

Catalog No. AC218360010
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC218360010 1 kg Plastic bottle
AC218365000 500 g Plastic bottle
AC218360050 5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC218360010 Supplier Thermo Scientific Chemicals Supplier No. 218360010
Only null left
Add to Cart

CAS: 631-61-8 | C2H7NO2 | 77.083 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer for analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 631-61-8
Molecular Formula C2H7NO2
Molecular Weight (g/mol) 77.083
MDL Number MFCD00013066
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
Synonym ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
PubChem CID 517165
ChEBI CHEBI:62947
IUPAC Name azanium;acetate
SMILES CC(=O)[O-].[NH4+]

Specifications

pH 6.5 to 7.5 (5% soln. at 20°C)
Melting Point 114.0°C
CAS Min % 98.0
Color White
CAS Max % 100.0
Assay Percent Range 98+%
Linear Formula CH3CO2NH4
Beilstein 02, 96
Fieser 01,38; 07,11
Merck Index 15, 492
Solubility Information Solubility in water: 1480g/L (20°C). Other solubilities: soluble in alcohol, slightly soluble in acetone, in methanol: 7.89 g/100mL (15°C)
Formula Weight 77.08
Percent Purity ≥98%
Grade Analytical
Quantity 1 kg
Flash Point 136°C
Infrared Spectrum Authentic
Packaging Plastic bottle
Physical Form Crystals or Crystalline Powder
Chemical Name or Material Acetic acid, ammonium salt, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.