Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, 99.5%, for analysis

Catalog No. AC258560025
Change view
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC258560025 2.5 L Glass Bottle
AC258560010 1 L Glass Bottle
AC258560250 25 L Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC258560025 Supplier Thermo Scientific Chemicals Supplier No. 258560025
  • $304.00
    $200.65 / Each

    Save $103.35

    34% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $935.00
    $617.65 / Case of 4 EA

    Save $317.35

    34% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 75-05-8 | C2H3N | 41.053 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -46.0°C
CAS Min % 99.4
CAS Max % 100.0
Density 0.7810g/mL
Quantity 2.5 L
Flash Point 2°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3430 to 1.3450
Residue after Evaporation 0.005% max.
Specific Gravity 0.781
Assay Percent Range 99.5%
Linear Formula CH3CN
Viscosity 0.36 mPa.s (20°C)
Water 0.1% max. (Coulometric)
Beilstein 02, 183
Fieser 02,13; 12,450; 13,156; 15,1
Merck Index 15, 69
Boiling Point 81.0°C to 82.0°C
Solubility Information Solubility in water: miscible. Other solubilities: soluble in alcohol, ethyl acetate, acetone, ether
Formula Weight 41.05
Percent Purity 99.5%
Grade Analytical
Chemical Name or Material Acetonitrile
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.