Learn More
Acetonitrile, 99.8%, for HPLC
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -46.0°C |
| Absorbance | (1cm cell vs HPLC−grade water), 0.005 max. at 260nm, 0.005 max. at 270nm, 0.005 max. at 280nm, 0.01 max. at 250nm, 0.015 max. at 240nm, 0.05 max. at 230nm |
| Acidity | 0.0008 meq/g max. |
| Decomposition Information | >500°C |
| Density | 0.7810g/mL |
| Boiling Point | 81.0°C to 82.0°C |
| Quantity | 2.5 L |
| Flash Point | 2°C |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Causes serious eye irritation. Harmful if inhaled. Harmful if swallowed. Harmful in contact with skin. Highly flammable liquid and vapor. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment. |
| EINECSNumber | 200-835-2 |
| RTECSNumber | AL7700000 |
| TSCA | TSCA |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.