Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Petroleum ether, extra pure, boiling range 60-95°C

Catalog No. AC292510010
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC292510010 1 L Glass Bottle
AC292510025 2.5 L Glass Bottle
AC292510100 10 L Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC292510010 Supplier Thermo Scientific Chemicals Supplier No. 292510010
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 64742-49-0 | C6H14 | 86.178 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64742-49-0
Molecular Formula C6H14
Molecular Weight (g/mol) 86.178
MDL Number MFCD00081849
InChI Key AFABGHUZZDYHJO-UHFFFAOYSA-N
Synonym isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane
PubChem CID 7892
SMILES CCCC(C)C

Specifications

Melting Point -40.0°C
Density 0.7000g/mL
Quantity 1 L
Flash Point −26°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3880 to 1.3920 (20°C, 589nm)
Residue after Evaporation 0.005% max.
Specific Gravity 0.7
Viscosity 0.57 mm2/s (25°C)
Boiling Point 60.0°C to 95.0°C
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with absolute alcohol, benzene,, chloroform, ether, carbon disulfide, carbon, tetrachloride and oils except castor oil.
IUPAC Name 2-methylpentane
Grade Extra Pure
Chemical Name or Material Petroleum ether
Show More Show Less
Hazard Category Danger
Hazard Statement GHS P Statement
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.