Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl 1,3-acetonedicarboxylate, 95%

Catalog No. AC325630500
Change view
Click to view available options
Quantity:
50 mL
250 mL
Packaging:
Glass bottle
Catalog No. Quantity Packaging
AC325630500 50 mL Glass bottle
AC325632500 250 mL Glass bottle
2 options

Catalog No. AC325630500

Supplier: Thermo Scientific Chemicals 325630500

Only null left
Add to Cart

CAS: 105-50-0 | C9H14O5 | 202.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-50-0
Molecular Formula C9H14O5
Molecular Weight (g/mol) 202.21
MDL Number MFCD00009200
InChI Key ZSANYRMTSBBUCA-UHFFFAOYSA-N
Synonym diethyl 1,3-acetonedicarboxylate, diethyl 3-oxoglutarate, 1,5-diethyl 3-oxopentanedioate, pentanedioic acid, 3-oxo-, diethyl ester, ethyl acetonedicarboxylate, ethyl 3-oxoglutarate, diethyl acetone-1,3-dicarboxylate, diethyl acetone dicarboxylate, 3-oxopentanedioic acid diethyl ester, acetonedicarboxylic acid diethyl ester
PubChem CID 66045
IUPAC Name diethyl 3-oxopentanedioate
SMILES CCOC(=O)CC(=O)CC(=O)OCC

Specifications

Color Colorless to Yellow
Density 1.1120g/mL
Boiling Point 250.0°C
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4380 to 1.4420
Linear Formula C2H5O2CCH2COCH2CO2C2H5
Quantity 50 mL
Beilstein 03, 791
Specific Gravity 1.112
Solubility Information Solubility in water: 10g/L in water (20°C)
Formula Weight 202.21
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Diethyl 1, 3-acetonedicarboxylate, 95%
Show More Show Less
Hazard Category MOISTURE SENSITIVE
EINECSNumber 203-302-2
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.