Learn More
Acetonitrile, 99.9%, for HPLC gradient grade, ACS reagent, meets the requirem. of Reag. Ph.Eur.

Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -46.0°C |
| Density | 0.7810g/mL |
| Quantity | 2.5 L |
| Flash Point | 2°C |
| Packaging | Glass Bottle |
| Refractive Index | 1.3430 to 1.3450 |
| Specific Gravity | 0.781 |
| Assay Percent Range | 99.9% |
| Linear Formula | CH3CN |
| Viscosity | 0.36 mPa.s (20°C) |
| Show More |
Safety and Handling
| EINECSNumber | 200-835-2 |
| RTECSNumber | AL7700000 |
| TSCA | TSCA |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.