Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, 99.9%, Extra Dry, AcroSeal™

Catalog No. AC326950025
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC326950025 2.5 L Glass Bottle
AC326951000 100 mL AcroSeal™ Glass Bottle
AC326950010 1 L AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC326950025 Supplier Thermo Scientific Chemicals Supplier No. 326950025
Only null left
Add to Cart

Methanol, 99.9%, CH4O, CAS Number-67-56-1, methylol, methyl alcohol, colonial spirit, carbinol, wood alcohol, wood naphtha, pyroxylic spirit, methyl hydroxide, wood spirit, columbian spirit, 1L, 0.002% max. (as HCOOH), 0.0005 meq/g max., 64.7 deg.C, CHEBI:17790, Colorless, 0.7910g/mL, 200-659-6 | CAS: 67-56-1 | CH4O | 32.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98.0°C
Color Colorless
Acidity 0.002% max. (as HCOOH)
Density 0.7910g/mL
Quantity 2.5 L
Flash Point 12°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3280 to 1.3300
Residue after Evaporation 0.0005 % max.
Assay Percent Range 99.9%
Specific Gravity 0.791
Linear Formula CH3OH
Viscosity 0.55 mPa.s (20°C)
Water 0.005% Max. (Coulometric)
Solubility Information Solubility in water: miscible with water. Other solubilities: miscible with ethanol, ether, benzene, ketones and, most other organic solvents
Boiling Point 64.7°C
Formula Weight 32.04
Percent Purity 99.9%
Grade Water <50ppm
Chemical Name or Material Methanol
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-659-6
RTECSNumber PC1400000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.