Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, 99.9%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals

Catalog Number AC326951000
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Glass Bottle
67-56-1
CH4O
32.04
MFCD00004595
OKKJLVBELUTLKV-UHFFFAOYSA-N
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
887
CHEBI:17790
methanol
CO
67-56-1
CH4O
32.04
MFCD00004595
OKKJLVBELUTLKV-UHFFFAOYSA-N
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
887
CHEBI:17790
methanol
CO

Methanol, 99.9%, CH4O, CAS Number-67-56-1, methylol, methyl alcohol, colonial spirit, carbinol, wood alcohol, wood naphtha, pyroxylic spirit, methyl hydroxide, wood spirit, columbian spirit, 1L, 0.002% max. (as HCOOH), 0.0005 meq/g max., 64.7 deg.C, CHEBI:17790, Colorless, 0.7910g/mL, 200-659-6

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -98.0°C
Color Colorless
Acidity 0.002% max. (as HCOOH)
Density 0.7910g/mL
Boiling Point 64.7°C
Quantity 100 mL
Flash Point 12°C
Infrared Spectrum Authentic
Packaging AcroSeal™ Glass Bottle
Physical Form Liquid
Refractive Index 1.3280 to 1.3300
Residue after Evaporation 0.0005 % max.
Assay Percent Range 99.9%
Specific Gravity 0.791
Linear Formula CH3OH
Viscosity 0.55 mPa.s (20°C)
Water 0.005% Max. (Coulometric)
Solubility Information Solubility in water: miscible with water. Other solubilities: miscible with ethanol, ether, benzene, ketones and, most other organic solvents
Formula Weight 32.04
Percent Purity 99.9%
Grade Water <50ppm
Chemical Name or Material Methanol
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-659-6
RTECSNumber PC1400000
TSCA TSCA

RUO – Research Use Only

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit