Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Petroleum ether, Extra Dry, boiling range 40-60°C, water <50ppm, AcroSeal™

Catalog No. AC396921000
Change view
Click to view available options
Quantity:
100 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC396921000 100 mL
AC396920010 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC396921000 Supplier Thermo Scientific Chemicals Supplier No. 396921000
  • $105.00
    $67.65 / Each of 1

    Save $37.35

    36% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 64742-49-0 | C6H14 | 86.178 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64742-49-0
Molecular Formula C6H14
Molecular Weight (g/mol) 86.178
MDL Number MFCD00081849
InChI Key AFABGHUZZDYHJO-UHFFFAOYSA-N
Synonym isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane
PubChem CID 7892
IUPAC Name Petroleum ether
SMILES CCCC(C)C

Specifications

Color Colorless
Density 0.6500g/mL
Quantity 100 mL
Flash Point −45°C
Physical Form Liquid
Refractive Index 1.3660 to 1.3710
Residue after Evaporation 0.001% max.
Specific Gravity 0.65
Viscosity 0.45 mm2/s (25°C)
Water 0.005% Max. (Coulometric)
Boiling Point 40.0°C to 65.0°C
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with absolute alcohol, benzene,, chloroform, ether, carbon disulfide, carbon, tetrachloride and oils except castor oil
Grade Extra Dry
Chemical Name or Material Petroleum ether
Show More Show Less
Hazard Category Danger
Hazard Statement GHS P Statement
Highly flammable liquid and vapour.
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Do not breathe dust/fume/gas/mist/vapors/spray.
Do not get in eyes, on skin, or on clothing.
Avoid release to the environment.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.