Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
Calcium hydroxide, 95+%, ACS reagent
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                1 kg
                            
                        
                            
                                2.5 kg
                            
                        
                    
                            
                                Packaging:
                            
                        
                        
                            
                                Plastic bottle
                            
                        
                    Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
            Specifications
Specifications
| Boiling Point | 2850.0°C | 
| Melting Point | 580.0°C | 
| Color | White | 
| Physical Form | Amorphous Powder | 
| Quantity | 1 kg | 
| Assay Percent Range | 95+% | 
| Linear Formula | Ca(OH)2 | 
| Merck Index | 15, 1675 | 
| Solubility Information | Solubility in water: 1.65g/L (20°C). Other solubilities: soluble in acids (evolution of heat) and nh4cl sol, soluble in glycerol and sugar, insoluble in alcohol | 
| Formula Weight | 74.09 | 
| Show More | 
Safety and Handling
| Hazard Category | GHS Signal Word: Danger | 
| Hazard Statement | GHS H Statement Causes serious eye damage.  | 
                                
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Immediately  | 
                                
| EINECSNumber | 215-137-3 | 
RUO – Research Use Only
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction