Learn More
1,4-Dioxane, 99+%, ACS reagent, stabilized, meets the requirements of Reag.Ph.Eur.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 12.0°C |
| CAS Min % | 99.0 |
| CAS Max % | 100.0 |
| Density | 1.0340g/mL |
| Quantity | 1 L |
| Flash Point | 12°C |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Refractive Index | 1.4205 to 1.4220 |
| Residue after Evaporation | 0.005% max. |
| Show More |
Safety and Handling
| Hazard Category | SIGNAL WORD: DANGER! |
| Hazard Statement | GHS H Statement: Causes serious eye irritation. Suspected of causing cancer. May cause respiratory irritation. Highly flammable liquid and vapor. May form explosive peroxides. Repeated exposure may cause skin dryness or cracking. |
| Precautionary Statement | Emergency Overview Flammable liquid and vapor. May form explosive peroxides. Possible cancer hazard. May cause cancer based on animal data. Irritating to eyes, respiratory system and skin. Inhalation may cause central nervous system effects. Repeated exposure may cause skin dryness or cracking. Hygroscopic. NFPA Health: 2 Flammability: 3 Instability: 1 |
| EINECSNumber | 204-661-8 |
| RTECSNumber | JG8225000 |
| TSCA | TSCA |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.