Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
n-Pentane, 95%, technical grade
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                5 L
                            
                        
                            
                                25 L
                            
                        
                    
                            
                                Packaging:
                            
                        
                        
                            
                                Metal Drum
                            
                        
                    Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
            Specifications
Specifications
| Melting Point | -130.0°C | 
| Color | Colorless | 
| CAS Min % | 94.0 | 
| CAS Max % | 100.0 | 
| Density | 0.6260g/mL | 
| Boiling Point | 36.0°C | 
| Quantity | 25 L | 
| Flash Point | −49°C | 
| Infrared Spectrum | Authentic | 
| Packaging | Metal Drum | 
| Show More | 
Safety and Handling
| Hazard Category | Danger | 
| Hazard Statement | GHS P Statement May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor. Repeated exposure may cause skin dryness or cracking. | 
| Precautionary Statement | GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment. | 
RUO – Research Use Only
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction