Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylenes, technical

Catalog No. AC444240050
Change view
Click to view available options
Quantity:
5 L
Packaging:
Plastic Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC444240050 5 L Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC444240050 Supplier Thermo Scientific Chemicals Supplier No. 444240050
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 1330-20-7 | C8H10 | 106.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1330-20-7
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
InChI Key IVSZLXZYQVIEFR-UHFFFAOYSA-N
IUPAC Name 1,3-xylene
SMILES CC1=CC(C)=CC=C1

Specifications

Melting Point -34.0°C
Density 0.8650g/mL
Quantity 5 L
Flash Point 23°C
Packaging Plastic Drum
Refractive Index 1.4940 to 1.4980
Specific Gravity 0.865
Linear Formula C6H4(CH3)2
Viscosity 0.6 mPa.s (20°C)
Merck Index 15,10278
Boiling Point 136.0°C to 140.0°C
Solubility Information Solubility in water: 0.1g/L water (20°C). Other solubilities: soluble in methanol,ethanol,2-propanol,dmf,,methylene chloride,acetone and ether
Formula Weight 106.17
Grade Technical
Chemical Name or Material Xylenes
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 215-535-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.