Learn More
n-Hexane, 97+%, for HPLC, ACS reagent, meets the requirm. of Reag. Ph.Eur., Thermo Scientific Chemicals

Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -95.0°C |
| Density | 0.6590g/mL |
| Boiling Point | 69.0°C |
| Flash Point | -22°C |
| Assay Percent Range | 97% min. (GC) |
| Packaging | Glass Bottle |
| Refractive Index | 1.3748 to 1.3810 |
| Linear Formula | CH3(CH2)4CH3 |
| Quantity | 1 L |
| Merck Index | 15,4732 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Causes skin irritation. May cause damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long las |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Use personal protective equipment as required. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or d |
| EINECSNumber | 203-777-6 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.