Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metaphosphoric acid, ACS reagent, stabilized

Catalog No. AC450250025
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass Bottle

CAS: 37267-86-0 | HO3P | 79.979 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37267-86-0, 50813-16-6
Molecular Formula HO3P
Molecular Weight (g/mol) 79.979
InChI Key UEZVMMHDMIWARA-UHFFFAOYSA-N
Synonym metaphosphoric acid, meta-phosphoric acid, hydrogenphosphonate, m-phosphoric acid, phosphenic acid, unii-mtk99r3uv0, metaphosphoric acid hpo3, x-phosphate-group, phosphonate 1-, hydrogen phosphonate
PubChem CID 3084658
SMILES OP(=O)=O

Specifications

Packaging Glass Bottle
Quantity 2.5 kg
Linear Formula (HPO3)n
Solubility Information Solubility in water: soluble. Other solubilities: slowly forming the ortho-acid, soluble in alcohol
Grade ACS Reagent
Chemical Name or Material Metaphosphoric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.