Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphoric acid, for HPLC

Catalog No. AC463861000
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC463861000 100 mL
AC463865000 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC463861000 Supplier Thermo Scientific Chemicals Supplier No. 463861000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 7664-38-2 | H3O4P | 97.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.99
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
IUPAC Name phosphoric acid
SMILES OP(O)(O)=O

Specifications

Melting Point 42.0°C
Density 1.7000 g/mL
Specific Gravity 1.7
Quantity 100 mL
Linear Formula H3PO4
Solubility Information Solubility aq. soln.: soluble. Other solubilities: soluble ethanol
Formula Weight 98
Grade HPLC
Chemical Name or Material Phosphoric acid
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Causes severe skin burns and eye damage.

May be corrosive to metals.

Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

EINECSNumber 231-633-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.