Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boric Acid (Crystalline/Electrophoresis), Fisher BioReagents

Catalog No. BP1681 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Glass Bottle
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP168-1 1 kg Poly Bottle
BP168-500 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP168-1
Only null left
Add to Cart
This high-purity electrophoresis-grade reagent is used in the preparation of Tris/Borate/EDTA electrophoresis buffers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

pH 3.8 to 4.8
Color White
Melting Point 169°C
Heavy Metals (as Pb) 0.001% max.
Identification Pass Test
Non_volatile Matter 0.05% max.
Packaging Poly Bottle
Vapor Pressure 2.7mbar at 20°C
Assay Percent Range ≥99 %
Quantity 1 kg
Physical Form Powder/Solid
Merck Index 15, 1339
Formula Weight 61.83
Percent Purity ≥99%
Grade DNase-Free
Chemical Name or Material Boric Acid
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
May impair fertility. Irritating to eyes and skin. May cause irritation of respiratory tract. May cause central nervous system effects. Ingestion may cause gastrointestinal irritation, nausea, vomiting and diarrhea. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:2
Flammability:0
Instability:1
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.