Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ampicillin Sodium Salt (Crystalline Powder), Fisher BioReagents™

Catalog No. BP176025 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP1760-25 25 g Poly Bottle
BP1760-5 5 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. BP1760-25
Only null left
Add to Cart
This item is not returnable. View return policy

For selection for ampicillin resistance in mutated and transformed cells | CAS: 69-52-3 | C16H21N3NaO4S | 374.411 g/mol

For selection for ampicillin resistance in mutated and transformed cells

For selection for ampicillin resistance in mutated and transformed cells. Antimicrobial spectrum: Gram-negative and Gram-positive bacteria.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-52-3
Molecular Formula C16H21N3NaO4S
Molecular Weight (g/mol) 374.411
InChI Key BSFVNXCYXDYHOD-ZQDFAFASSA-N
Synonym Ampicillin
PubChem CID 131673879
IUPAC Name (2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium
SMILES [HH].CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)C.[Na]

Specifications

Type Ampicillin Sodium Salt
Physical Form Solid
pH 8.0 to 10.0
Melting Point 215°C
Packaging Poly Bottle
Potency Inclusive between 845μg/mg
Water 2% Max.
Color White
Quantity 25 g
Merck Index 15, 583
Formula Weight 371.39
Chemical Name or Material Ampicillin Sodium Salt
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
May cause allergic respiratory and skin reaction. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:2
Flammability:0
Instability:0
Recommended Storage 4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.