Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Arginine (White Crystals or Crystalline Powder), Fisher BioReagents

Catalog No. BP370100 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP370-100 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP370-100

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This amino acid is suitable for use in tissue culture systems requiring additives.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74-79-3
Molecular Formula C6H14N4O2
Molecular Weight (g/mol) 174.20
MDL Number MFCD00002635
InChI Key ODKSFYDXXFIFQN-UHFFFAOYNA-N
Synonym l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l
PubChem CID 6322
ChEBI CHEBI:16467
IUPAC Name (2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid
SMILES NC(CCCN=C(N)N)C(O)=O

Specifications

Melting Point 223°C
Ammonium 0.02% max.
Arsenic (As) 1ppm max.
pH 11.4
Color White
Heavy Metals (as Pb) 10ppm max.
Identification Pass Test
Loss on Drying 0.5% max.
Assay Percent Range 98.5 to 101.0 %
Other Amino Acids Not Detected
Packaging Glass Bottle
Residue after Ignition 0.1% max.
Specific Rotation 26.9 to 27.9° (+ or -)
Merck Index 15, 770
State of Solution (%T) 98% min.
Alpha Vector Arginine
Quantity 100 g
Formula Weight 174.2
Percent Purity 98.5 to 101.0%
Physical Form Solid
Chemical Name or Material L-Arginine
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.