Learn More
Specifications
Specifications
| Melting Point | -63°C |
| Color | Colorless |
| Decomposition Information | 290°C |
| Density | 1.480g/cm³ |
| Quantity | 4 L |
| Identification | Pass Test |
| Packaging | Aluminum Can |
| Physical Form | Liquid |
| Assay Percent Range | ≥99.8 % |
| Residue after Evaporation | 0.001% max. |
| Show More |
Safety and Handling
| Hazard Statement | WARNING! |
| Precautionary Statement | Emergency Overview Possible cancer hazard. May cause cancer based on animal data. Irritating to eyes and skin. May cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. May be harmful if inhaled. May cause irritation of respiratory tract. Danger of serious damage to health by prolonged exposure. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. NFPA Health:2 Flammability:1 Instability:0 |
| DOTInformation | DOT Class 6.1, : Poison |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.