Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Ether Anhydrous (Certified ACS), Fisher Chemical™

Catalog No. E1381 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Aluminum Can
Steel Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
E138-1 1 L Aluminum Can
E138-500 500 mL Aluminum Can
E138-4 4 L Aluminum Can
E138-20 20 L Steel Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. E138-1
Only null left
Add to Cart

Meet or exceeds the latest ACS specifications. Actual lot of analysis on label

Stabilized with BHT as preservative. Meet or exceeds the latest ACS specifications. Actual lot of analysis on label

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Melting Point -116°C
Color Colorless
Density 0.714g/cm³
Quantity 1 L
Identification Pass Test
Packaging Aluminum Can
Physical Form Liquid
Residue after Evaporation 0.001% max.
Assay Percent Range ≥99 %
Titratable Acid 0.0002mEq/g max.
Linear Formula C2H5OC2H5
Vapor Pressure 587mbar at 20°C
Viscosity 0.2448 cP at 20°C
UN Number 1155
Water 0.03% max.
Boiling Point 34.6°C
Formula Weight 74.12
Percent Purity ≥99%
Grade ACS
Chemical Name or Material Ethyl Ether Anhydrous
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Extremely flammable liquid and vapor. Vapor may cause flash fire. Harmful if swallowed. Irritating to eyes and skin. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. May cause irritation of respiratory tract. Repeated exposure may cause skin dryness or cracking. Hygroscopic. Use personal protective equipment. Keep away from open flames
DOTInformation DOT Class 3, : Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.