Learn More
Specifications
Specifications
| Melting Point | -116°C |
| Absorbance | 0.01 AU max. at 400nm, 0.03 AU max. at 270nm, 0.1 AU max. at 254nm, 0.2 AU max. at 242nm, 0.4 AU max. at 231nm, 0.6 AU max. at 224nm, 0.8 AU max. at 221nm, 1 AU max. at 220nm |
| Color | Colorless |
| Acidity | 5ppm max. |
| Density | 0.714g/cm³ |
| Quantity | 4 L |
| Flash Point | -45°C (-113°F) |
| Identification | Pass Test |
| Packaging | Tin Can |
| Physical Form | Liquid |
| Show More |
Safety and Handling
| Hazard Statement | DANGER! |
| Precautionary Statement | Emergency Overview Extremely flammable liquid and vapor. Vapor may cause flash fire. Harmful if swallowed. Irritating to eyes and skin. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. May cause irritation of respiratory tract. Repeated exposure may cause skin dryness or cracking. Hygroscopic. Wear personal protective equipment. Handle under an inert atmosphere. Ensure adequate ventilation. Avoid contact with skin, eyes and clothing. Avoid contact with skin, eyes and clothing. Do not breathe vapors or spray mist. NFPA Health:1 Flammability:4 Instability:1 |
| DOTInformation | DOT Class 3, : Flammable Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.