Learn More
Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Melting Point | -116°C |
| Absorbance | 0.01 AU max. at 300nm, 0.01 AU max. at 350nm, 0.02 AU max. at 270nm, 0.08 AU max. at 250nm, 1.00 AU max. at 218nm |
| Color | Colorless |
| Density | 0.714g/cm³ |
| Quantity | 4 L |
| Packaging | Amber Glass |
| Physical Form | Liquid |
| Residue after Evaporation | 3ppm max. |
| Assay Percent Range | ≥99.5 % |
| Vapor Pressure | 587mbar at 20°C |
| Show More |
Safety and Handling
| Hazard Statement | DANGER! |
| Precautionary Statement | Emergency Overview Extremely flammable liquid and vapor. Vapor may cause flash fire. Harmful if swallowed. Irritating to eyes and skin. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. May cause irritation of respiratory tract. Repeated exposure may cause skin dryness or cracking. Hygroscopic. Wear personal protective equipment. Handle under an inert atmosphere. Ensure adequate ventilation. Avoid contact with skin, eyes and clothing. Avoid contact with skin, eyes and clothing. Do not breathe vapors or spray mist. NFPA Health:1 Flammability:4 Instability:1 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
