Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mobile phase for HPLC, Acetonitrile with formic acid (0.1%), HPLC Grade, Fisher Chemical

Catalog No. HB98234 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
HB9823-4 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. HB9823-4
Get 1 Extra Chemical When You Buy 2
View all promotions
Add to Cart
  • Ideal mobile phase blend developed for use with HPLC instruments
  • Pre-mixed for ease-of-use
  • Formic acid is a common mobile phase additive used in HPLC separations
  • Formic acid (FA) controls the pH of the mobile phase by functioning as a buffer
  • FA is also known to improve the peak shapes of the resulting separation
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.05
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name formic acid
SMILES C(=O)O

Specifications

Density 0.7810g/cm³
Quantity 4 L
Flash Point 6°C (43°F)
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Assay Percent Range ≥98.5 %
Grade HPLC
Chemical Name or Material Acetonitrile with 0.1% Formic Acid (v/v)
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Irritating to eyes. May cause skin and respiratory tract irritation. May cause central nervous system effects. Liver and kidney injuries may occur. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.