Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methyl iso-Butyl Ketone (Certified ACS), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
1 L
4 L
20 L
200 L
Packaging:
Glass Bottle
Steel Drum
Steel Pail
Specifications
Specifications
Melting Point | -84°C |
Density | 0.80g/cm³ |
Boiling Point | 117°C |
Quantity | 20 L |
Identification | Pass Test |
Packaging | Steel Pail |
Physical Form | Liquid |
Assay Percent Range | ≥98.5 % |
Residue after Evaporation | 0.005% max. |
Titratable Acid | 0.002mEq/g max. |
Show More |
Safety and Handling


Hazard Statement | DANGER! |
Precautionary Statement | Emergency Overview Flammable liquid and vapor. Harmful by inhalation. Irritating to eyes, respiratory system and skin. May cause central nervous system effects. Repeated exposure may cause skin dryness or cracking. Aspiration hazard if swallowed - can enter lungs and cause damage. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. If peroxide formation is suspected, do not open or move container. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. NFPA Health:2 Flammability:3 Instability:1 |
DOTInformation | DOT Class 3, : Flammable Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction